missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichloropyrazine, TRC
Click to view available options
Quantity:
250 g
Unit Size:
250g
Specifications
Specifications
| Chemical Name or Material | 2,6-Dichloropyrazine |
| CAS | 4774-14-5 |
| Molecular Formula | C4 H2 Cl2 N2 |
| Quantity | 250 g |
| InChI Formula | InChI=1S/C4H2Cl2N2/c5-3-1-7-2-4(6)8-3/h1-2H |
| SMILES | Clc1cncc(Cl)n1 |
| IUPAC Name | 2,6-dichloropyrazine |
| Molecular Weight (g/mol) | 148.98 |
| Formula Weight | 147.9595 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction