missing translation for 'onlineSavingsMsg'
Learn More

(2,6-Dichlorophenylthio)acetic acid, 99%

Product Code. 11419420
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419420 1 g 1g
11429420 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419420 Supplier Thermo Scientific Alfa Aesar Supplier No. H50282.03
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 21248-45-3 | C8H5Cl2O2S | 236.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21248-45-3
Molecular Formula C8H5Cl2O2S
Molecular Weight (g/mol) 236.09
MDL Number MFCD00052108
InChI Key URNRRSJGOOTQBO-UHFFFAOYSA-M
Synonym 2,6-dichlorophenylthio acetic acid, 2-2,6-dichlorophenyl thio acetic acid, 2,6-dichlorophenyl thio acetic acid, 2,6-dichlorophenyl sulfanyl acetic acid, 2-2,6-dichlorophenyl sulfanylacetic acid, acmc-1cg2g, 2,6-dichlorothiophenoxyacetic acid, 2-2,6-dichlorophenylthio acetic acid, 2-2,6-dichlorophenyl sulfanyl acetic acid
PubChem CID 1810591
IUPAC Name 2-(2,6-dichlorophenyl)sulfanylacetic acid
SMILES [O-]C(=O)CSC1=C(Cl)C=CC=C1Cl

Specifications

Quantity 1 g
Formula Weight 237.11
Percent Purity 99%
Chemical Name or Material (2,6-Dichlorophenylthio)acetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.