missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenylhydrazine hydrochloride, 97%, Thermo Scientific™

Product Code. 10113164
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10113164 25 g Amber Glass Bottle 25g
10411854 1 g Amber Glass Bottle 1g
10503754 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10113164 Supplier Thermo Scientific Maybridge Supplier No. BTB08663EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 50709-36-9
Molecular Formula C6H7Cl3N2
Molecular Weight (g/mol) 213.486
InChI Key CQNIYLLTIOPFCJ-UHFFFAOYSA-N
Synonym 2,6-dichlorophenylhydrazine hydrochloride, 2,6-dichlorophenyl hydrazine hydrochloride, 1-2,6-dichlorophenyl hydrazine hydrochloride, 2,6-dichlorophenyl hydrazine monohydrochloride, hydrazine, 2,6-dichlorophenyl-, monohydrochloride, 2,6-dichlorophenylhydrazinehydrochloride, acmc-209kpe, ksc491e8j, 2,6-dichlorophenylhydrazine, chloride, 2,6-dichlorophenylhydrazine hcl
PubChem CID 2723909
IUPAC Name (2,6-dichlorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=C(C(=C1)Cl)NN)Cl.Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula Cl2C6H3NHNH2·HCl
Quantity 25 g
Formula Weight 213.49
Percent Purity 97%
Chemical Name or Material 1-(2, 6-Dichlorophenyl)hydrazine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.