missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenylacetonitrile, 98%, Thermo Scientific™

Product Code. 11428867
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428867 500 g 500g
11408867 25 g 25g
11418867 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428867 Supplier Thermo Scientific Alfa Aesar Supplier No. A19983.36

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 3215-64-3
Molecular Formula C8H5Cl2N
Molecular Weight (g/mol) 186.035
MDL Number MFCD00001901
InChI Key AOEJUUCUKRUCEF-UHFFFAOYSA-N
Synonym 2,6-dichlorophenylacetonitrile, 2,6-dichlorobenzyl cyanide, 2-2,6-dichlorophenyl acetonitrile, 2,6-dichlorophenyl acetonitrile, 2,6-dichlorobenzeneacetonitrile, benzeneacetonitrile, 2,6-dichloro, acetonitrile, 2,6-dichlorophenyl, 2,6-dichloro-phenyl-acetonitrile, 2-2,6-dichlorophenyl ethanenitrile
PubChem CID 76678
IUPAC Name 2-(2,6-dichlorophenyl)acetonitrile
SMILES C1=CC(=C(C(=C1)Cl)CC#N)Cl

Especificaciones

Melting Point 74°C to 77°C
Odor Characteristic
Quantity 500 g
UN Number UN3439
Beilstein 1910482
Formula Weight 186.04
Percent Purity 98%
Chemical Name or Material 2,6-Dichlorophenylacetonitrile
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.