missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenylacetic Acid, TRC

Product Code. 30005264
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005264 100 g 100g
30007507 25 g 25g
30005988 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005264 Supplier Toronto Research Chemicals Supplier No. TRCD436305100G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6575-24-2
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.04
Synonym 2,6-Dichlorobenzeneacetic Acid, 2-(2,6-Dichlorophenyl)Acetic Acid
SMILES OC(=O)Cc1c(Cl)cccc1Cl

Specifications

Quantity 100 g
InChI Formula InChI=1S/C8H6Cl2O2/c9-6-2-1-3-7(10)5(6)4-8(11)12/h1-3H,4H2,(H,11,12)
Formula Weight 203.9745
Chemical Name or Material 2,6-Dichlorophenylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.