missing translation for 'onlineSavingsMsg'
Learn More

(2,6-Dichlorophenyl)methanamine, TRC

Product Code. 30377081
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30377081 2.5 g 2.5g
30372392 1 g -
30376068 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30377081 Supplier Toronto Research Chemicals Supplier No. TRCD4378402.5G
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6575-27-5
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.04
Synonym 2,6-Dichlorobenzylamine, 2,6-Dichlorobenzenemethanamine
SMILES NCc1c(Cl)cccc1Cl

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C7H7Cl2N/c8-6-2-1-3-7(9)5(6)4-10/h1-3H,4,10H2
Formula Weight 174.9956
Chemical Name or Material (2,6-Dichlorophenyl)methanamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.