missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenolindophenol Sodium Salt Hydrate, Honeywell Fluka™

Product Code. 15613910 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15613910 5 g Glass Bottle 5g
15603910 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15613910 Supplier Honeywell-Fluka Supplier No. 331255G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Redox indicator

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-45-1
Molecular Formula C12H6Cl2NNaO2
Molecular Weight (g/mol) 290.07
MDL Number MFCD00150014
InChI Key FHLDWQLHDYCXKI-UHFFFAOYSA-M
PubChem CID 23697355
ChEBI CHEBI:948
SMILES [Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1

Specifications

Melting Point >300°C
UN Number NONH for all modes of transport
Beilstein 3641229
IUPAC Name sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate
Formula Weight 290.08g/mol (anhydrous basis)
Loss on Drying ≤14% (120°C)
Odor Odourless
Packaging Glass Bottle
Color Green
Quantity 5 g
Physical Form Powder
Chemical Name or Material 2,6-Dichlorophenolindophenol Sodium Salt Hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.