missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenethyl alcohol, 96%

Product Code. 10028831 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10028831 1 g Glass bottle 1g
10222322 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10028831 Supplier Thermo Scientific Acros Supplier No. 350760010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 30595-79-0 | C8H8Cl2O | 191.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30595-79-0
Molecular Formula C8H8Cl2O
Molecular Weight (g/mol) 191.05
MDL Number MFCD00800674
InChI Key ZBQPKQUIKJDGIX-UHFFFAOYSA-N
Synonym 2,6-dichlorophenethyl alcohol, 2-2,6-dichlorophenyl ethanol, 2,6-dichlorophenethylalcohol, 2-2,6-dichlorophenyl ethan-1-ol, 2,6-dichlorophenethanol, benzeneethanol, 2,6-dichloro, acmc-209hha, 2,6-dichlorobenzeneethanol, ksc493m0h, benzeneethanol,2,6-dichloro
PubChem CID 2734100
IUPAC Name 2-(2,6-dichlorophenyl)ethanol
SMILES OCCC1=C(Cl)C=CC=C1Cl

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 59°C to 65°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 191.06
Percent Purity 96%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material 2, 6-Dichlorophenethyl alcohol, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.