missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloronicotinic acid, 98+%, Thermo Scientific Chemicals

Catalog Number 10571334
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
38496-18-3
C6H3Cl2NO2
192
MFCD00075583
AJPKQSSFYHPYMH-UHFFFAOYSA-N
2,6-dichloronicotinic acid, 2,6-dichloro-nicotinic acid, 2,6-dichloro-3-pyridinecarboxylic acid, dichloronicotinic acid, 2,6-dichloronicotininc acid, 3-pyridinecarboxylic acid, 2,6-dichloro, 2,6-dichloronicotinincacid, pubchem1225, 2,6-dichlornicotinsaure, acmc-209j0g
854047
2,6-dichloropyridine-3-carboxylic acid
C1=CC(=NC(=C1C(=O)O)Cl)Cl
This item is not returnable. View return policy
38496-18-3
C6H3Cl2NO2
192
MFCD00075583
AJPKQSSFYHPYMH-UHFFFAOYSA-N
2,6-dichloronicotinic acid, 2,6-dichloro-nicotinic acid, 2,6-dichloro-3-pyridinecarboxylic acid, dichloronicotinic acid, 2,6-dichloronicotininc acid, 3-pyridinecarboxylic acid, 2,6-dichloro, 2,6-dichloronicotinincacid, pubchem1225, 2,6-dichlornicotinsaure, acmc-209j0g
854047
2,6-dichloropyridine-3-carboxylic acid
C1=CC(=NC(=C1C(=O)O)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 147°C to 150°C
Color White or Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 192
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 2, 6-Dichloronicotinic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Dichloronicotinic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'