missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloronicotinic acid, 97%

Product Code. 11324386
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324386 5 g 5g
11314386 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11324386 Supplier Thermo Scientific Alfa Aesar Supplier No. L20002.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 38496-18-3 | C6H3Cl2NO2 | 191.995 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38496-18-3
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 191.995
MDL Number MFCD00075583
InChI Key AJPKQSSFYHPYMH-UHFFFAOYSA-N
Synonym 2,6-dichloronicotinic acid, 2,6-dichloro-nicotinic acid, 2,6-dichloro-3-pyridinecarboxylic acid, dichloronicotinic acid, 2,6-dichloronicotininc acid, 3-pyridinecarboxylic acid, 2,6-dichloro, 2,6-dichloronicotinincacid, pubchem1225, 2,6-dichlornicotinsaure, acmc-209j0g
PubChem CID 854047
IUPAC Name 2,6-dichloropyridine-3-carboxylic acid
SMILES C1=CC(=NC(=C1C(=O)O)Cl)Cl

Specifications

Melting Point 146°C to 151°C
Quantity 5 g
Beilstein 136114
Formula Weight 192
Percent Purity 97%
Chemical Name or Material 2,6-Dichloronicotinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.