missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichloroindophenol sodium salt, MP Biomedicals™
Click to view available options
Quantity:
1 g
Specifications
Specifications
Color | Blue-Green |
Quantity | 1 g |
Formula Weight | 290.08 |
Physical Form | Crystalline Powder |
Chemical Name or Material | 2, 6-Dichloroindophenol sodium salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'