missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloroindophenol sodium salt hydrate, Honeywell Fluka™

Product Code. 15635120 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15635120 1 g 1g
15655120 5 g 5g
15645120 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15635120 Supplier Honeywell-Fluka Supplier No. 361801G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

≥97.0% (calc. based on dry substance, AT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-45-1
Molecular Formula C12H6Cl2NNaO2
Molecular Weight (g/mol) 290.07
MDL Number MFCD00150014
InChI Key FHLDWQLHDYCXKI-UHFFFAOYSA-M
Synonym 2,6-dichloroindophenol sodium salt, 2,6-dichlorophenolindophenol sodium salt, tillman's reagent, tillman's reagenz, dichlorphenol-indophenolnatrium, 2,6-dichloroindophenol sodium, unii-kad7q8xo1y, sodium 2,6-dichloroindophenol, sodium 2,6-dichloroindophenolate, 2,6-dichloroindophenol, sodium salt
PubChem CID 23697355
ChEBI CHEBI:948
IUPAC Name sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate
SMILES [Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1

Specifications

Quantity 1 g
UN Number NONH for all modes of transport
Beilstein 3641229
Formula Weight 290.08g/mol (anhydrous basis)
Percent Purity ≥97% (calculated based on dry substance, AT)
Chemical Name or Material 2,6-Dichloroindophenol sodium salt hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.