missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloroindophenol, sodium salt hydrate, 95%, Thermo Scientific Chemicals

Product Code. 10636932
Click to view available options
Quantity:
1 g
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1266615-56-8
Molecular Formula C12H6Cl2NNaO2
Molecular Weight (g/mol) 290.07
MDL Number MFCD00150014
InChI Key CVSUAFOWIXUYQA-UHFFFAOYSA-M
Synonym Tillman's reagent hydrate
PubChem CID 23696612
SMILES [Na+].[O-]C1=CC=C(C=C1)N=C1C=C(Cl)C(=O)C(Cl)=C1

Specifications

Assay Percent Range 98+%
Solubility Information Solubility in water: soluble. Other solubilities: freely soluble in alcohol
Infrared Spectrum Authentic
IUPAC Name sodium 4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate
Formula Weight 290.07
Loss on Drying 14% max. (1 g, 120°C)
Percent Purity ≥98%
Packaging Glass bottle
Suitability For determination of ascorbic acid: Passes Test
Color Dark Green
Quantity 10 g
Physical Form Powder
Chemical Name or Material 2, 6-Dichloroindophenol, sodium salt hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'