missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloroindophenol sodium salt hydrate

Product Code. 11459342
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459342 25 g 25g
11449342 5 g 5g
11469342 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459342 Supplier Thermo Scientific Alfa Aesar Supplier No. A10107.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1266615-56-8 | C12H6Cl2NNaO2 | 290.07 g/mol

2,6-Dichloroindophenol sodium salt hydrate is suitable for vitamin C determination. It acts as a redox dye to investigate the efficiency of gold nanoparticles (Au-NP) for the enzymatic activity of glucose oxidase. It is also used to compose UVB specific dosimeter and to measure the rate of photosynthesis. It serves as a good reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichloroindophenol sodium salt hydrate is suitable for vitamin C determination. It acts as a redox dye to investigate the efficiency of gold nanoparticles (Au-NP) for the enzymatic activity of glucose oxidase. It is also used to compose UVB specific dosimeter and to measure the rate of photosynthesis. It serves as a good reducing agent.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1266615-56-8
Molecular Formula C12H6Cl2NNaO2
Molecular Weight (g/mol) 290.07
MDL Number MFCD00150014
InChI Key FHLDWQLHDYCXKI-UHFFFAOYSA-M
Synonym 2,6-dichloroindophenol sodium salt, 2,6-dichlorophenolindophenol sodium salt, tillman's reagent, tillman's reagenz, dichlorphenol-indophenolnatrium, 2,6-dichloroindophenol sodium, unii-kad7q8xo1y, sodium 2,6-dichloroindophenol, sodium 2,6-dichloroindophenolate, 2,6-dichloroindophenol, sodium salt
PubChem CID 23697355
ChEBI CHEBI:948
IUPAC Name sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate
SMILES [Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 3641229
Sensitivity Light sensitive
Merck Index 14,3068
Solubility Information Soluble in water.
Formula Weight 290.08 (Anhydrous)
Chemical Name or Material 2,6-Dichloroindophenol sodium salt hydrate
compliance-icons
Product Identifier
  • 2,6-Dichloroindophenol sodium salt hydrate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.