missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichloroindophenol sodium salt hydrate, Thermo Scientific Chemicals
26.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
2,6-Dichloroindophenol sodium salt hydrate is suitable for vitamin C determination. It acts as a redox dye to investigate the efficiency of gold nanoparticles (Au-NP) for the enzymatic activity of glucose oxidase. It is also used to compose UVB specific dosimeter and to measure the rate of photosynthesis. It serves as a good reducing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-Dichloroindophenol sodium salt hydrate is suitable for vitamin C determination. It acts as a redox dye to investigate the efficiency of gold nanoparticles (Au-NP) for the enzymatic activity of glucose oxidase. It is also used to compose UVB specific dosimeter and to measure the rate of photosynthesis. It serves as a good reducing agent.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
2,6-Dichloroindophenol sodium salt hydrate is suitable for vitamin C determination. It acts as a redox dye to investigate the efficiency of gold nanoparticles (Au-NP) for the enzymatic activity of glucose oxidase. It is also used to compose UVB specific dosimeter and to measure the rate of photosynthesis. It serves as a good reducing agent.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | >300°C |
Quantity | 5 g |
Beilstein | 3641229 |
Sensitivity | Light sensitive |
Merck Index | 14,3068 |
Solubility Information | Soluble in water. |
Formula Weight | 290.08 (Anhydrous) |
Chemical Name or Material | 2,6-Dichloroindophenol sodium salt hydrate |
Safety and Handling
Product Identifier
- 2,6-Dichloroindophenol sodium salt hydrate
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'