missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloroindophenol Sodium Salt Dihydrate, TRC

Product Code. 17943415
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17943415 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17943415 Supplier Toronto Research Chemicals Supplier No. TRCD4359081G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1082681-24-0
Molecular Formula C12H6Cl2NNaO2 . 2[H2O]
Quantity 1 g
Synonym [p-[(3,5-Dichloro-4-oxo-2,5-cyclohexadien-1-ylidene)amino]phenoxy]- (7CI), 2,6-
InChI Formula InChI=1S/C12H7Cl2NO2.Na.H2O/c13-10-5-8(6-11(14)12(10)17)15-7-1-3-9(16)4-2-7;;/h1-6,16H;;1H2
SMILES [Na].O=C1C(Cl)=CC(=NC2=CC=C(O)C=C2)C=C1Cl.O
IUPAC Name 2,6-dichloro-4-((4-hydroxyphenyl)imino)cyclohexa-2,5-dien-1-one, sodium salt hydrate
Molecular Weight (g/mol) 326.12
Formula Weight 324.99
Chemical Name or Material 2,6-Dichloroindophenol Sodium Salt Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.