missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzylamine, 98%

Product Code. 10415282 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10415282 5 g Glass bottle 5g
10313392 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10415282 Supplier Thermo Scientific Acros Supplier No. 350740050
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6575-27-5 | C7H7Cl2N | 176.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6575-27-5
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.04
MDL Number MFCD00047928
InChI Key VLVLNNQMURDGPM-UHFFFAOYSA-N
Synonym 2,6-dichlorobenzylamine, 2,6-dichlorophenyl methanamine, benzenemethanamine, 2,6-dichloro, 2,6-dichlorophenyl methylamine, 1-2,6-dichlorophenyl methanamine, pubchem15690, 2,6-dichloro-benzylamine, acmc-20ao98, chembl13305, benzenemethanamine,2,6-dichloro
PubChem CID 485432
IUPAC Name (2,6-dichlorophenyl)methanamine
SMILES NCC1=C(Cl)C=CC=C1Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5570 to 1.559
Quantity 5 g
Beilstein 12, III, 2347
Formula Weight 176.05
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 6-Dichlorobenzylamine
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Dichlorobenzylamine
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.