missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzoyl chloride, 98%

Product Code. 11472607
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472607 50 g 50g
11462607 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11472607 Supplier Thermo Scientific Alfa Aesar Supplier No. A14839.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4659-45-4 | C7H3Cl3O | 209.45 g/mol

2,6-Dichlorobenzoyl chloride acts as an important intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is employed in the substrate activity screening method for the rapid development of novel substrates and their conversion into non-peptidic inhibitors of Cys and Ser proteases. It is also used in the preparation of 1-acyliridoles and (-)-aspicilin. In addition to this, it undergoes esterification of (fluoren-9-ylmethoxy)carbonyl (Fmoc)-amino acids to 4-alkoxybenzyl alcohol polystyrene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichlorobenzoyl chloride acts as an important intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is employed in the substrate activity screening method for the rapid development of novel substrates and their conversion into non-peptidic inhibitors of Cys and Ser proteases. It is also used in the preparation of 1-acyliridoles and (-)-aspicilin. In addition to this, it undergoes esterification of (fluoren-9-ylmethoxy)carbonyl (Fmoc)-amino acids to 4-alkoxybenzyl alcohol polystyrene.

Solubility
Miscible with alcohol, ether and acetone. Slightly miscible with heptane. Immiscible with water.

Notes
Moisture sensitive. Incompatible with alcohols, oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4659-45-4
Molecular Formula C7H3Cl3O
Molecular Weight (g/mol) 209.45
MDL Number MFCD00000662
InChI Key JBLIDPPHFGWTKU-UHFFFAOYSA-N
Synonym benzoyl chloride, 2,6-dichloro, 2,6-dichlorobenzoylchloride, unii-394f6s81wy, 2,6-dichloro-benzoyl chloride, ccris 8617, pubchem18674, acmc-209k7i, 2.6-dichlorobenzoylchloride, dsstox_cid_12891, dsstox_rid_79043
PubChem CID 78392
IUPAC Name 2,6-dichlorobenzoyl chloride
SMILES C1=CC(=C(C(=C1)Cl)C(=O)Cl)Cl

Specifications

Melting Point 15°C to 18°C
Density 1.463
Boiling Point 130°C to 132°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.56
Quantity 50 g
UN Number UN3265
Beilstein 639531
Sensitivity Moisture sensitive
Solubility Information Miscible with alcohol,ether and acetone. Slightly miscible with heptane. Immiscible with water.
Formula Weight 209.46
Percent Purity 98%
Chemical Name or Material 2,6-Dichlorobenzoyl chloride
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.