missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzonitrile, 98%

Product Code. 11427207
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11427207 100 g 100g
11464060 10 g 10g
11417207 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11427207 Supplier Thermo Scientific Alfa Aesar Supplier No. A14986.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1194-65-6 | C7H3Cl2N | 172.008 g/mol

2,6-dichlorobenzonitrile in which Q-amino-G- chlorobenzonitrile is used as starting material. And it is used for the synthesis of diclofenac sodium oxacillin and guanabenz acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-dichlorobenzonitrile in which Q-amino-G- chlorobenzonitrile is used as starting material. And it is used for the synthesis of diclofenac sodium oxacillin and guanabenz acetate.

Solubility
Soluble in water (0.018g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1194-65-6
Molecular Formula C7H3Cl2N
Molecular Weight (g/mol) 172.008
MDL Number MFCD00001781
InChI Key YOYAIZYFCNQIRF-UHFFFAOYSA-N
Synonym dichlobenil, dichlobanil, casoron, prefix d, benzonitrile, 2,6-dichloro, casaron, du-sprex, decabane, surfassol, casoron 133
PubChem CID 3031
ChEBI CHEBI:943
IUPAC Name 2,6-dichlorobenzonitrile
SMILES C1=CC(=C(C(=C1)Cl)C#N)Cl

Specifications

Melting Point 143°C to 147°C
Boiling Point 270°C
Odor Aromatic
Quantity 100 g
UN Number UN3077
Beilstein 1909167
Merck Index 14,3042
Solubility Information Soluble in water (0.018g/L).
Formula Weight 172.01
Percent Purity 98%
Chemical Name or Material 2,6-Dichlorobenzonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.