missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzene-1-sulfonyl chloride, ≥95%, Thermo Scientific™

Product Code. 10059551
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10059551 10 g 10g
10581123 1 g 1g
10367404 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10059551 Supplier Thermo Scientific Maybridge Supplier No. TL00344EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 6579-54-0
Molecular Formula C6H3Cl3O2S
Molecular Weight (g/mol) 245.498
MDL Number MFCD00052311
InChI Key WGGKQIKICKLWGN-UHFFFAOYSA-N
Synonym 2,6-dichlorobenzene-1-sulfonyl chloride, 2,6-dichlorobenzenesulfonylchloride, 2,6-dichlorophenylsulfonyl chloride, 2,6-dichlorobenzenesulphonyl chloride, unii-8523kf52zn, benzenesulfonyl chloride, 2,6-dichloro, 2,6-dichloro-benzenesulfonyl chloride, 2,6-dichlorobenzene sulfonyl chloride, pubchem5117, acmc-209ns8
PubChem CID 522183
IUPAC Name 2,6-dichlorobenzenesulfonyl chloride
SMILES C1=CC(=C(C(=C1)Cl)S(=O)(=O)Cl)Cl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula Cl2C6H3SO2Cl
Quantity 10 g
Formula Weight 245.51
Percent Purity 95%
Chemical Name or Material 2, 6-Dichlorobenzene-1-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.