missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzamide, 98%

Product Code. 11494203
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494203 100 g 100g
11474203 5 g 5g
11484203 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494203 Supplier Thermo Scientific Alfa Aesar Supplier No. A10865.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2008-58-4 | C7H5Cl2NO | 190.023 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2008-58-4
Molecular Formula C7H5Cl2NO
Molecular Weight (g/mol) 190.023
MDL Number MFCD00007975
InChI Key JHSPCUHPSIUQRB-UHFFFAOYSA-N
Synonym benzamide, 2,6-dichloro, 2,6-bam, unii-e9jwf529eb, 2,6-dichlorobenzoic acid amide, bam, e9jwf529eb, benzamide,6-dichloro, dsstox_cid_2170, acmc-209f5j, dsstox_rid_76511
PubChem CID 16183
ChEBI CHEBI:28435
IUPAC Name 2,6-dichlorobenzamide
SMILES C1=CC(=C(C(=C1)Cl)C(=O)N)Cl

Specifications

Melting Point 198°C to 200°C
Quantity 100 g
Beilstein 1869103
Formula Weight 190.03
Percent Purity 98%
Chemical Name or Material 2,6-Dichlorobenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.