missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichlorobenzaldoxime, 97%, Thermo Scientific Chemicals
£20.50 - £164.00
Chemical Identifiers
CAS | 25185-95-9 |
---|---|
Molecular Formula | C7H5Cl2NO |
Molecular Weight (g/mol) | 190.02 |
MDL Number | MFCD00013938 |
InChI Key | YBSXDWIAUZOFFV-ONNFQVAWSA-N |
Synonym | 2,6-dichlorobenzaldoxime, 2,6-dichlorobenzaldehyde oxime, 2,6-dichlorobenzaloxime, 2,6-dichloro benzyloxime, z-2,6-dichlorobenzaldehyde oxime, 2,6-dichlorophenyl hydroxyimino methane, z-n-2,6-dichlorophenyl methylidene hydroxylamine, z-2,6-dichlorobenzaldehydeoxime, e-n-2,6-dichlorophenyl methylidene hydroxylamine, e-2,6-dichlorobenzaldehyde oxime |
PubChem CID | 9581041 |
IUPAC Name | (NE)-N-[(2,6-dichlorophenyl)methylidene]hydroxylamine |
SMILES | O\N=C\C1=C(Cl)C=CC=C1Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11484060
|
Thermo Scientific Alfa Aesar
A11351.06 |
5 g |
£20.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470143
|
Thermo Scientific Alfa Aesar
A11351.14 |
25 g |
£55.60
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480143
|
Thermo Scientific Alfa Aesar
A11351.22 |
100 g |
£164.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
25185-95-9 | |
190.02 | |
YBSXDWIAUZOFFV-ONNFQVAWSA-N | |
9581041 | |
O\N=C\C1=C(Cl)C=CC=C1Cl |
C7H5Cl2NO | |
MFCD00013938 | |
2,6-dichlorobenzaldoxime, 2,6-dichlorobenzaldehyde oxime, 2,6-dichlorobenzaloxime, 2,6-dichloro benzyloxime, z-2,6-dichlorobenzaldehyde oxime, 2,6-dichlorophenyl hydroxyimino methane, z-n-2,6-dichlorophenyl methylidene hydroxylamine, z-2,6-dichlorobenzaldehydeoxime, e-n-2,6-dichlorophenyl methylidene hydroxylamine, e-2,6-dichlorobenzaldehyde oxime | |
(NE)-N-[(2,6-dichlorophenyl)methylidene]hydroxylamine |
Specifications
25185-95-9 | |
C7H5Cl2NO | |
2046662 | |
YBSXDWIAUZOFFV-ONNFQVAWSA-N | |
(NE)-N-[(2,6-dichlorophenyl)methylidene]hydroxylamine | |
9581041 | |
97% |
147°C to 151°C | |
MFCD00013938 | |
2,6-dichlorobenzaldoxime, 2,6-dichlorobenzaldehyde oxime, 2,6-dichlorobenzaloxime, 2,6-dichloro benzyloxime, z-2,6-dichlorobenzaldehyde oxime, 2,6-dichlorophenyl hydroxyimino methane, z-n-2,6-dichlorophenyl methylidene hydroxylamine, z-2,6-dichlorobenzaldehydeoxime, e-n-2,6-dichlorophenyl methylidene hydroxylamine, e-2,6-dichlorobenzaldehyde oxime | |
O\N=C\C1=C(Cl)C=CC=C1Cl | |
190.02 | |
190.03 | |
2,6-Dichlorobenzaldoxime |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 246-720-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only