missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzaldoxime, 97%

Product Code. 11480143
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480143 100 g 100g
11484060 5 g 5g
11470143 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480143 Supplier Thermo Scientific Alfa Aesar Supplier No. A11351.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25185-95-9 | C7H5Cl2NO | 190.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25185-95-9
Molecular Formula C7H5Cl2NO
Molecular Weight (g/mol) 190.02
MDL Number MFCD00013938
InChI Key YBSXDWIAUZOFFV-ONNFQVAWSA-N
Synonym 2,6-dichlorobenzaldoxime, 2,6-dichlorobenzaldehyde oxime, 2,6-dichlorobenzaloxime, 2,6-dichloro benzyloxime, z-2,6-dichlorobenzaldehyde oxime, 2,6-dichlorophenyl hydroxyimino methane, z-n-2,6-dichlorophenyl methylidene hydroxylamine, z-2,6-dichlorobenzaldehydeoxime, e-n-2,6-dichlorophenyl methylidene hydroxylamine, e-2,6-dichlorobenzaldehyde oxime
PubChem CID 9581041
IUPAC Name (NE)-N-[(2,6-dichlorophenyl)methylidene]hydroxylamine
SMILES O\N=C\C1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 147°C to 151°C
Quantity 100 g
Beilstein 2046662
Formula Weight 190.03
Percent Purity 97%
Chemical Name or Material 2,6-Dichlorobenzaldoxime

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.