missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzaldehyde, 97+%

Product Code. 11470873
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11470873

Brand: Thermo Scientific Alfa Aesar A13295.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 83-38-5 | C7H4Cl2O | 175.008 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-38-5
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.008
MDL Number MFCD00003307
InChI Key DMIYKWPEFRFTPY-UHFFFAOYSA-N
Synonym benzaldehyde, 2,6-dichloro, ccris 6014, 2,6-dichloro benzaldehyde, 2,6-dichloro-benzaldehyde, 2,6-dichloro-benzaldehyd, 2,6-dichlorbenzaldehyd, pubchem8225, acmc-1bkey, 2,6dichloro-benzaldehyde, benzaldehyde,6-dichloro
PubChem CID 6737
IUPAC Name 2,6-dichlorobenzaldehyde
SMILES C1=CC(=C(C(=C1)Cl)C=O)Cl

Specifications

Melting Point 69°C to 73°C
Boiling Point 124°C (10 mmHg)
Flash Point 135°C (275°F)
Quantity 500 g
UN Number UN3261
Beilstein 386477
Sensitivity Air Sensitive
Formula Weight 175.01
Percent Purity 98%
Chemical Name or Material 2,6-Dichlorobenzaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.