missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloroaniline, TRC

Product Code. 30137468
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137468 50 g 50g
17929639 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137468 Supplier Toronto Research Chemicals Supplier No. TRCD43180550G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-31-1
Molecular Formula C6 H5 Cl2 N
Molecular Weight (g/mol) 162.02
Synonym 2,6-Dichloroaniline, Clonidine Hydrochloride Imp. C (EP), 2,6-Dichlorophenylamine, 2,6-Dichlorobenzenamine
IUPAC Name 2,6-dichloroaniline
SMILES Nc1c(Cl)cccc1Cl

Specifications

Quantity 50 g
InChI Formula InChI=1S/C6H5Cl2N/c7-4-2-1-3-5(8)6(4)9/h1-3H,9H2
Formula Weight 160.9799
Chemical Name or Material 2,6-Dichloroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.