missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichloroaniline, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
2,6-Dichloroaniline
Specifications
Specifications
| Chemical Name or Material | 2,6-Dichloroaniline |
| CAS | 608-31-1 |
| Analyte or Component Names | 2,6-Dichloroaniline |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H5 Cl2 N |
| Certifications/Compliance | ISO 17025 |
| Synonym | Clonidine Hydrochloride Imp. C (EP), Clonidine Imp. C (EP), 2,6-Dichloroaniline, Clonidine Hydrochloride Impurity C, Clonidine Impurity C |
| InChI Formula | InChI=1S/C6H5Cl2N/c7-4-2-1-3-5(8)6(4)9/h1-3H,9H2 |
| SMILES | Nc1c(Cl)cccc1Cl |
| IUPAC Name | 2,6-dichloroaniline |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction