missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichloroaniline, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
2,6-Dichloroaniline is used in the pharmaceutical industry for the synthesis of antibacterial agents of lomefloxacin, Lee uric acid and clonidine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-Dichloroaniline is used in the pharmaceutical industry for the synthesis of antibacterial agents of lomefloxacin, Lee uric acid and clonidine.
Solubility
Soluble in water (1.6 g/L at 20°C), dimethyl sulfoxide and methanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
2,6-Dichloroaniline is used in the pharmaceutical industry for the synthesis of antibacterial agents of lomefloxacin, Lee uric acid and clonidine.
Solubility
Soluble in water (1.6 g/L at 20°C), dimethyl sulfoxide and methanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Melting Point | 37°C to 40°C |
Boiling Point | 97°C (5 mmHg) |
Flash Point | 118°C (244°F) |
Quantity | 25 g |
UN Number | UN3442 |
Beilstein | 775084 |
Formula Weight | 162.02 |
Percent Purity | 98% |
Chemical Name or Material | 2,6-Dichloroaniline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'