missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Dichloroacetophenone, 98%

Product Code. 11312486
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312486 5 g 5g
11302486 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312486 Supplier Thermo Scientific Alfa Aesar Supplier No. L06348.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2040-05-3 | C8H6Cl2O | 189.04 g/mol

2',6'-Dichloroacetophenone is is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,6′-Dichloroacetophenone is is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2040-05-3
MDL Number MFCD00016339

Specifications

Melting Point 41°C to 44°C
Boiling Point 176°C to 180°C (83 mmHg)
Molecular Formula C8H6Cl2O
Quantity 5 g
Beilstein 2440104
Synonym 2',6'-dichloroacetophenone, 1-2,6-dichlorophenyl ethanone, 2,6-dichloroacetophenone, 1-2,6-dichlorophenyl ethan-1-one, ethanone, 1-2,6-dichlorophenyl, 2,6-dichloro acetophenone, 1-acetyl-2,6-dichlorobenzene, zlchem 1084, pubchem12362, acmc-1cobf
Solubility Information Insoluble in water.
InChI Key HYBDSXBLGCQKRE-UHFFFAOYSA-N
SMILES CC(=O)C1=C(C=CC=C1Cl)Cl
IUPAC Name 1-(2,6-dichlorophenyl)ethanone
Molecular Weight (g/mol) 189.04
PubChem CID 74877
Formula Weight 189.04
Percent Purity 98%
Chemical Name or Material 2',6'-Dichloroacetophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.