missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Dichloroacetophenone, 98%

Product Code. 11302486
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302486 1 g 1g
11312486 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302486 Supplier Thermo Scientific Alfa Aesar Supplier No. L06348.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2040-05-3 | C8H6Cl2O | 189.04 g/mol

2',6'-Dichloroacetophenone is is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,6′-Dichloroacetophenone is is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2040-05-3
MDL Number MFCD00016339

Specifications

Melting Point 41°C to 44°C
Boiling Point 176°C to 180°C (83 mmHg)
Molecular Formula C11H14O
Quantity 1 g
Beilstein 2440104
Synonym 2',4',5'-trimethylacetophenone, 1-2,4,5-trimethylphenyl ethanone, 1-2,4,5-trimethylphenyl ethan-1-one, acetophenone, 2',4',5'-trimethyl, ethanone, 1-2,4,5-trimethylphenyl, 1-acetyl-2,4,5-trimethylbenzene, acmc-20aofe, 2',5'-trimethylacetophenone, 2,4,5-trimethyl-acetophenone, acetophenone,4',5'-trimethyl
Solubility Information Insoluble in water.
InChI Key GENBEGZNCBFHSU-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1C(=O)C)C)C
IUPAC Name 1-(2,4,5-trimethylphenyl)ethanone
Molecular Weight (g/mol) 162.23
PubChem CID 74878
Formula Weight 189.04
Percent Purity 98%
Chemical Name or Material 2',6'-Dichloroacetophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.