missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-7-deazapurine, TRC

Product Code. 30369080
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369080 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30369080 Supplier Toronto Research Chemicals Supplier No. TRCD4340255G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,6-Dichloro-7-deazapurine
CAS 90213-66-4
Molecular Formula C6 H3 Cl2 N3
Quantity 5 g
Synonym 7H-Pyrrolo[2,3-d]pyrimidine, 2,4-dichloro- (ACI), 1H-Pyrrolo[2,3-d]pyrimidine, 2,4-dichloro- (9CI), 2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine (ACI), 2,4-Dichloro-1H-pyrrolo[2,3-d]pyrimidine
InChI Formula InChI=1S/C6H3Cl2N3/c7-4-3-1-2-9-5(3)11-6(8)10-4/h1-2H,(H,9,10,11)
SMILES Clc1nc(Cl)c2cc[nH]c2n1
IUPAC Name 2,4-dichloro-7H-pyrrolo[2,3-d]pyrimidine
Molecular Weight (g/mol) 188.01
Formula Weight 186.9704
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.