missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-5-fluoronicotinic acid, 97%, Thermo Scientific™

Product Code. 10728825
Change view
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass Bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10728825 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10728825 Supplier Thermo Scientific Maybridge Supplier No. MO07724EB
 Restricted Item
Estimated Shipment: 16-11-2026
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Formats
Denna artikel kan inte returneras. Se returpolicy

Specifikationer

CAS 82671-06-5
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H2Cl2FNO2
Quantity 25 g
Synonym 2,6-dichloro-5-fluoronicotinic acid, 2,6-dichloro-5-fluoro-nicotinic acid, 2,6-dichloro-5-fluoro-3-pyridinecarboxylic acid, 2,6-dichloro-5-fluoro-pyridine-3-carboxylic acid, 2,6-dichloro-5-fluoro nicotinic acid, 3-pyridinecarboxylic acid, 2,6-dichloro-5-fluoro, pubchem10943, acmc-1bluq, ksc448a5j, 2,6-dichloro-5-fluoro-nicotinicacid
InChI Key LTDGKGCHRNNCAC-UHFFFAOYSA-N
SMILES C1=C(C(=NC(=C1F)Cl)Cl)C(=O)O
IUPAC Name 2,6-dichloro-5-fluoropyridine-3-carboxylic acid
Molecular Weight (g/mol) 209.985
PubChem CID 2733659
Formula Weight 209.99
Percent Purity 97%
Chemical Name or Material 2, 6-Dichloro-5-fluoronicotinic acid
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.