missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-(trifluoromethyl)pyridin-3-amine, 97%, Thermo Scientific™

Product Code. 10715283
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10715283 1 g Amber Glass Bottle 1g
10149032 10 g Amber Glass Bottle 10g
10205624 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10715283 Supplier Thermo Scientific Maybridge Supplier No. SPB06360DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175277-67-5
Molecular Formula C6H3Cl2F3N2
Molecular Weight (g/mol) 231.00
MDL Number MFCD00173957
InChI Key BVJJNBRZQVLPIC-UHFFFAOYSA-N
PubChem CID 2735898
IUPAC Name 2,6-dichloro-4-(trifluoromethyl)pyridin-3-amine
SMILES NC1=C(Cl)N=C(Cl)C=C1C(F)(F)F

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 2,6-dichloro-4-trifluoromethyl pyridin-3-amine, 3-amino-2,6-dichloro-4-trifluoromethyl pyridine, 3-amino-2,6-dichloro-4-trifluoromethyl-pyridine, pubchem2205, 2,6-dichloro-3-amino-4-trifluoromethylpyridine, 2,6-dichloro-4-trifluoromethyl-3-pyridinamine, 2,6-dichloro-4-trifluoromethyl-3-pyridylamine, 2,6-dichloro-4-trifluoromethyl-pyridin-3-ylamine, 3-pyridinamine,2,6-dichloro-4-trifluoromethyl, 3-pyridinamine, 2,6-dichloro-4-trifluoromethyl
Formula Weight 231
Percent Purity 97%
Chemical Name or Material 2, 6-Dichloro-4-(trifluoromethyl)pyridin-3-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.