missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-(trifluoromethyl)nicotinonitrile, 97%, Thermo Scientific™

Product Code. 10140702
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10140702 10 g Amber Glass Bottle 10g
10744583 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10140702 Supplier Thermo Scientific Maybridge Supplier No. RF03850EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 13600-42-5
Molecular Formula C7HCl2F3N2
Molecular Weight (g/mol) 240.99
MDL Number MFCD00084977
InChI Key WRXXBTBGBXYHSG-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-trifluoromethyl nicotinonitrile, 3-cyano-2,6-dichloro-4-trifluoromethyl pyridine, 2,6-dichloro-3-cyano-4-trifluoromethylpyridine, 2,6-dichloro-4-trifluoromethyl pyridine-3-carbonitrile, 3-cyano-2,6-dichloro-4-trifluoromethyl-pyridine, 2,6-dichloro-4-trifluoromethyl-nicotinonitrile, 3-pyridinecarbonitrile, 2,6-dichloro-4-trifluoromethyl, pubchem9225, buttpark 4357-39, ksc496s7t
PubChem CID 2736726
IUPAC Name 2,6-dichloro-4-(trifluoromethyl)pyridine-3-carbonitrile
SMILES FC(F)(F)C1=CC(Cl)=NC(Cl)=C1C#N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 241
Percent Purity 97%
Chemical Name or Material 2, 6-Dichloro-4-(trifluoromethyl)nicotinonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.