missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-(trifluoromethyl)aniline, TRC

Product Code. 30381365
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381365 25 g
30383928 10 g
30375217 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381365 Supplier Toronto Research Chemicals Supplier No. TRCD43617025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24279-39-8
Molecular Formula C7 H4 Cl2 F3 N
Molecular Weight (g/mol) 230.014
Synonym Toluidine, 2,6-dichloro-α,α,α-trifluoro- (8CI), 2,6-Dichloro-4-(trifluoromethyl)benzenamine, 2,6-Dichloro-4-trifluoromethylaniline, 4-Amino-3,5-dichlorobenzotrifluoride
IUPAC Name 2,6-dichloro-4-(trifluoromethyl)aniline
SMILES Nc1c(Cl)cc(cc1Cl)C(F)(F)F

Specifications

Quantity 25 g
InChI Formula InChI=1S/C7H4Cl2F3N/c8-4-1-3(7(10,11)12)2-5(9)6(4)13/h1-2H,13H2
Formula Weight 228.9673
Chemical Name or Material 2,6-Dichloro-4-trifluoromethylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.