missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Product Code. 11465586
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465586 1 g 1g
11475586 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465586 Supplier Thermo Scientific Alfa Aesar Supplier No. B20728.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24279-39-8
Molecular Formula C7H4Cl2F3N
Molecular Weight (g/mol) 230.01
MDL Number MFCD00052918
InChI Key ITNMAZSPBLRJLU-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-trifluoromethyl aniline, 4-amino-3,5-dichlorobenzotrifluoride, 3,5-dichloro-4-amino benzotrifluoride, 3,5-dichloro-4-aminobenzotrifluoride, 2,6-dichloro-4-trifluoromethyl benzenamine, benzenamine, 2,6-dichloro-4-trifluoromethyl, 2,6-dichloro-4-trifluoromethyl phenylamine, 2,6-dichloro-4-trifluoromethyl-benzenamine, pubchem2187
PubChem CID 141094
IUPAC Name 2,6-dichloro-4-(trifluoromethyl)aniline
SMILES NC1=C(Cl)C=C(C=C1Cl)C(F)(F)F

Specifications

Melting Point 34°C to 37°C
Density 1.532
Boiling Point 60°C to 62°C (1 mmHg)
Flash Point 88°C (190°F)
Quantity 1 g
UN Number UN3077
Beilstein 2838819
Formula Weight 230.02
Percent Purity 97%
Chemical Name or Material 2,6-Dichloro-4-(trifluoromethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.