missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-(trifluoromethoxy)aniline, 95%, Thermo Scientific™

Product Code. 11455947
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455947 10 g 10g
11445947 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455947 Supplier Thermo Scientific Alfa Aesar Supplier No. B24482.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99479-66-0
Molecular Formula C7H4Cl2F3NO
Molecular Weight (g/mol) 246.01
MDL Number MFCD00190128
InChI Key FKISQWQHZULEEG-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-trifluoromethoxy aniline, benzenamine, 2,6-dichloro-4-trifluoromethoxy, 2,6-dichloro-4-trifluoromethoxy phenylamine, pubchem8461, acmc-209scs, 3.5-dichloro-4-amino trifluoromethoxybenzene, 2,6-dichloro-4-trifluoromethoxy-phenylamine, 3,5-dichloro-4-amino trifluoromethoxybenzene, 3,5-dichloro-4-aminotrifluoromethoxy benzene, 2,6-dichloro-4-trifluoromethoxy benzenamine
PubChem CID 688247
IUPAC Name 2,6-dichloro-4-(trifluoromethoxy)aniline
SMILES NC1=C(Cl)C=C(OC(F)(F)F)C=C1Cl

Specifications

Melting Point 31°C to 33°C
Flash Point >110°C (230°F)
Refractive Index 1.4981
Quantity 10 g
Formula Weight 246.02
Percent Purity 95%
Chemical Name or Material 2,6-Dichloro-4-(trifluoromethoxy)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.