missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-nitroaniline, 95%

Product Code. 11435697
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435697 500 g 500g
11494060 25 g 25g
11425697 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435697 Supplier Thermo Scientific Alfa Aesar Supplier No. B24283.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 99-30-9 | C6H4Cl2N2O2 | 207.01 g/mol

It is used as dyestuff, agricultural and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as dyestuff, agricultural and pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-30-9
Molecular Formula C6H4Cl2N2O2
Molecular Weight (g/mol) 207.01
MDL Number MFCD00007677
InChI Key BIXZHMJUSMUDOQ-UHFFFAOYSA-N
Synonym dichloran, dicloran, allisan, botran, dcna, ditranil, batran, bortran, resisan, dcna fungicide
PubChem CID 7430
ChEBI CHEBI:27864
IUPAC Name 2,6-dichloro-4-nitroaniline
SMILES C1=C(C=C(C(=C1Cl)N)Cl)[N+](=O)[O-]

Specifications

Melting Point 185°C to 192°C
Boiling Point 130°C (2 mmHg)
Odor Recognizable
Quantity 500 g
Beilstein 1459581
Solubility Information Insoluble in water.
Formula Weight 207.02
Percent Purity 95%
Chemical Name or Material 2,6-Dichloro-4-nitroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.