missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-(chloromethyl)pyridine, Thermo Scientific™

Product Code. 10141172
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10141172 1 g 1g
10497622 250 mg 250mg
10758645 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10141172 Supplier Thermo Scientific Maybridge Supplier No. MO00840DA
 Restricted Item
Estimated Shipment: 26-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 101990-72-1
Molecular Formula C6H4Cl3N
Molecular Weight (g/mol) 196.455
InChI Key UYBVOPLURVHJOX-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-chloromethyl pyridine, pyridine, 2,6-dichloro-4-chloromethyl, acmc-20dwvl, 2,6-dichloro-isonicotinyl chloride, 2,6-dichloro-4-chloromethyl-pyridine, 4-chloromethyl-2,6-dichloropyridine, pyridine, 2,6-dichloro-4-chloromethyl-9ci, pyridine,2,6-dichloro-4-chloromethyl
PubChem CID 2794626
IUPAC Name 2,6-dichloro-4-(chloromethyl)pyridine
SMILES C1=C(C=C(N=C1Cl)Cl)CCl

Specifications

Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 196.46
Percent Purity 95%
Chemical Name or Material 2, 6-Dichloro-4-(chloromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.