missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichloro-3-iodopyridine, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used as pharmaceutical raw material and intermediate.
Solubility
Slightly soluble in water.
Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
It is used as pharmaceutical raw material and intermediate.
Solubility
Slightly soluble in water.
Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 74°C to 79°C |
| Color | White to Brown |
| Quantity | 5 g |
| UN Number | UN2811 |
| Beilstein | 6135696 |
| Sensitivity | Light sensitive |
| Synonym | 2,6-dichloro-3-iodo-pyridine, pyridine, 2,6-dichloro-3-iodo, zlchem 993, acmc-20aihk, pubchem14239, tpc-py080, 2,6-dichloro-3-iodo pyridine, pyridine,2,6-dichloro-3-iodo, 2,6-dichloro-3-iodopyridine |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 273.88 |
| Percent Purity | 98% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction