Learn More
2,6-Dichloro-1,4-benzoquinone, 97+%
Description
2,6-Dichloro-1,4-benzoquinone is the suitable reagent used to in a study to evaluate the diffusion coefficients (D) for a family of quinones, nitroaromatics, ferrocenes and aromatic hydrocarbon compounds, in acetonitrile by single potential step chronoamperometry.1 It may be used in the preparation of 2,3,5-Trichloro-1,4-dihydroquinone, 2,3,5-trichloro-1,4-benzoquinone and 2,6-Dichloro-1,4-dihydroquinone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,6-Dichloro-1,4-benzoquinone is the suitable reagent used to in a study to evaluate the diffusion coefficients (D) for a family of quinones, nitroaromatics, ferrocenes and aromatic hydrocarbon compounds, in acetonitrile by single potential step chronoamperometry.1 It may be used in the preparation of 2,3,5-Trichloro-1,4-dihydroquinone, 2,3,5-trichloro-1,4-benzoquinone and 2,6-Dichloro-1,4-dihydroquinone.
Solubility
Insoluble in water. Solubility in methanol (almost transparency).
Notes
Store away from reducing agents and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 119°C to 121°C |
| Quantity | 25 g |
| Solubility Information | Insoluble in water. Solubility in methanol (almost transparency). |
| Formula Weight | 176.98 |
| Percent Purity | 98% |
| Chemical Name or Material | 2,6-Dichloro-1,4-benzoquinone |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.