missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-1,4-benzoquinone, 97+%

Product Code. 11461830
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11461830

Brand: Thermo Scientific Alfa Aesar H27542.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 697-91-6 | C6H2Cl2O2 | 176.98 g/mol

2,6-Dichloro-1,4-benzoquinone is the suitable reagent used to in a study to evaluate the diffusion coefficients (D) for a family of quinones, nitroaromatics, ferrocenes and aromatic hydrocarbon compounds, in acetonitrile by single potential step chronoamperometry.1 It may be used in the preparation of 2,3,5-Trichloro-1,4-dihydroquinone, 2,3,5-trichloro-1,4-benzoquinone and 2,6-Dichloro-1,4-dihydroquinone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichloro-1,4-benzoquinone is the suitable reagent used to in a study to evaluate the diffusion coefficients (D) for a family of quinones, nitroaromatics, ferrocenes and aromatic hydrocarbon compounds, in acetonitrile by single potential step chronoamperometry.1 It may be used in the preparation of 2,3,5-Trichloro-1,4-dihydroquinone, 2,3,5-trichloro-1,4-benzoquinone and 2,6-Dichloro-1,4-dihydroquinone.

Solubility
Insoluble in water. Solubility in methanol (almost transparency).

Notes
Store away from reducing agents and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 697-91-6
Molecular Formula C6H2Cl2O2
Molecular Weight (g/mol) 176.98
MDL Number MFCD00037159
InChI Key JCARTGJGWCGSSU-UHFFFAOYSA-N
Synonym 2,6-dichloro-1,4-benzoquinone, 2,6-dichloro-p-benzoquinone, 2,6-dichlorobenzoquinone, 2,6-dichloroquinone, 2,5-cyclohexadiene-1,4-dione, 2,6-dichloro, 2,6-dichloquinone, p-quinone, 2,6-dichloro, p-benzoquinone, 2,6-dichloro, 2,6-dichloro-2,5-cyclohexadiene-1,4-dione, p-quinone,6-dichloro
PubChem CID 12771
IUPAC Name 2,6-dichlorocyclohexa-2,5-diene-1,4-dione
SMILES C1=C(C(=O)C(=CC1=O)Cl)Cl

Specifications

Melting Point 119°C to 121°C
Quantity 5 g
Solubility Information Insoluble in water. Solubility in methanol (almost transparency).
Formula Weight 176.98
Percent Purity 98%
Chemical Name or Material 2,6-Dichloro-1,4-benzoquinone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.