missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromotoluene, 98+%

Product Code. 11493784
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493784 5 g 5g
11483784 1 g 1g
11403794 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493784 Supplier Thermo Scientific Alfa Aesar Supplier No. A11832.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 69321-60-4 | C7H6Br2 | 249.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69321-60-4
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.93
MDL Number MFCD00013524
InChI Key OCSKCBIGEMSDIS-UHFFFAOYSA-N
Synonym 2,6-dibromotoluene, benzene, 1,3-dibromo-2-methyl, 1,3-dibromo-2-methyl-benzene, pubchem3926, acmc-1bagq, ksc493g1t, #, 2,6-dibromotoluene 1g
PubChem CID 34681
IUPAC Name 1,3-dibromo-2-methylbenzene
SMILES CC1=C(Br)C=CC=C1Br

Specifications

Melting Point 2°C to 6°C
Density 1.812
Boiling Point 112°C to 113°C (7 mmHg)
Refractive Index 1.606
Quantity 5 g
Beilstein 3235502
Formula Weight 249.94
Percent Purity ≥98%
Chemical Name or Material 2,6-Dibromotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.