missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromotoluene, 98+%

Product Code. 11483784
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11483784

Brand: Thermo Scientific Alfa Aesar A11832.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 69321-60-4 | C7H6Br2 | 249.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69321-60-4
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.93
MDL Number MFCD00013524
InChI Key OCSKCBIGEMSDIS-UHFFFAOYSA-N
Synonym 2,6-dibromotoluene, benzene, 1,3-dibromo-2-methyl, 1,3-dibromo-2-methyl-benzene, pubchem3926, acmc-1bagq, ksc493g1t, #, 2,6-dibromotoluene 1g
PubChem CID 34681
IUPAC Name 1,3-dibromo-2-methylbenzene
SMILES CC1=C(Br)C=CC=C1Br

Specifications

Melting Point 2°C to 6°C
Density 1.812
Boiling Point 112°C to 113°C (7 mmHg)
Refractive Index 1.606
Quantity 1 g
Beilstein 3235502
Formula Weight 249.94
Percent Purity ≥98%
Chemical Name or Material 2,6-Dibromotoluene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.