missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dibromopyridine, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | 2,6-Dibromopyridine |
| CAS | 626-05-1 |
| Molecular Formula | C5H3Br2N |
| Quantity | 50 g |
| Synonym | NSC 613 |
| InChI Formula | InChI=1S/C5H3Br2N/c6-4-2-1-3-5(7)8-4/h1-3H |
| SMILES | Brc1cccc(Br)n1 |
| IUPAC Name | 2,6-dibromopyridine |
| Molecular Weight (g/mol) | 236.89 |
| Formula Weight | 234.8632 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction