missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromopyridine, 98%

Product Code. 10213950 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10213950 100 g Glass bottle 100g
10203950 25 g Glass bottle 25g
11916291 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10213950 Supplier Thermo Scientific Acros Supplier No. 112911000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 626-05-1 | C5H3Br2N | 236.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-05-1
Molecular Formula C5H3Br2N
Molecular Weight (g/mol) 236.89
InChI Key FEYDZHNIIMENOB-UHFFFAOYSA-N
PubChem CID 12274
IUPAC Name 2,6-dibromopyridine
SMILES C1=CC(=NC(=C1)Br)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 118.0°C to 119.0°C
Color Gray to White
Flash Point 213°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H3N)Br2
Quantity 100 g
Beilstein 20, V,5, 435
Synonym pyridine, 2,6-dibromo, 2,6-dibromo-pyridine, 2,6-dibromo pyridine, 2,6dibromopyridine, 2,6-dibrompyridine, 2,6 dibromopyridine, 2.6-dibromopyridine, pubchem3204, 2, 6-dibromopyridine, acmc-209n5v
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,ethanol,dioxane,chloroform,,carbon tetrachloride,heptane,aq. naoh,aq. h2so4
Formula Weight 236.89
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 6-Dibromopyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Dibromopyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.