missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromopyridine, 97%, Thermo Scientific™

Product Code. 10789395
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10789395 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10789395 Supplier Thermo Scientific Maybridge Supplier No. SB01233DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 626-05-1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C5H3Br2N
Linear Formula (C5H3N)Br2
MDL Number MFCD00006223
Quantity 1 g
Synonym pyridine, 2,6-dibromo, 2,6-dibromo-pyridine, 2,6-dibromo pyridine, 2,6dibromopyridine, 2,6-dibrompyridine, 2,6 dibromopyridine, 2.6-dibromopyridine, pubchem3204, 2, 6-dibromopyridine, acmc-209n5v
InChI Key FEYDZHNIIMENOB-UHFFFAOYSA-N
SMILES C1=CC(=NC(=C1)Br)Br
IUPAC Name 2,6-dibromopyridine
Molecular Weight (g/mol) 236.894
PubChem CID 12274
Formula Weight 236.89
Percent Purity 97%
Chemical Name or Material 2, 6-Dibromopyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.