missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromo-4-methylaniline, 98+%

Product Code. 11443407
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443407 100 g 100g
11433407 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11443407 Supplier Thermo Scientific Alfa Aesar Supplier No. A17184.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6968-24-7 | C7H7Br2N | 264.948 g/mol

2,6-Dibromo-4-methylaniline is used as coupling reagent in spectrophotometric determination of carbaryl in various environmental samples. It was also in preparation of 2-bromo-6-iodo-4-methylaniline. 2,6-Dibromo-4-methylaniline undergoes Suzuki-coupling reaction with 2-dihydroxyboryl-3-methyl-2-cyclopenten-1-one during synthesis of the dinuclear dichlorotitanium complexes. 2,6-Dibromo-4-methylaniline is also used as an intermediate for the synthesis of pharmaceutucals, agrochemicals, dyes and other organic chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dibromo-4-methylaniline is used as coupling reagent in spectrophotometric determination of carbaryl in various environmental samples. It was also in preparation of 2-bromo-6-iodo-4-methylaniline. 2,6-Dibromo-4-methylaniline undergoes Suzuki-coupling reaction with 2-dihydroxyboryl-3-methyl-2-cyclopenten-1-one during synthesis of the dinuclear dichlorotitanium complexes. 2,6-Dibromo-4-methylaniline is also used as an intermediate for the synthesis of pharmaceutucals, agrochemicals, dyes and other organic chemicals.

Solubility
Insoluble in water.

Notes
Stable under ordinary conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6968-24-7
Molecular Formula C7H7Br2N
Molecular Weight (g/mol) 264.948
MDL Number MFCD00007641
InChI Key ATDIROHVRVQMRO-UHFFFAOYSA-N
Synonym 2,6-dibromo-p-toluidine, 3,5-dibromo-4-aminotoluene, benzenamine, 2,6-dibromo-4-methyl, 2,6-dibromo-4-methyl aniline, 2,6-dibromo-4-methyl-phenylamine, 2,6-dibromo-4-methylphenylamine, 2,6-dibromo-4-methyl-benzenamine, pubchem2334, acmc-209oag, 2,6-dibromo-p-toludine
PubChem CID 81427
IUPAC Name 2,6-dibromo-4-methylaniline
SMILES CC1=CC(=C(C(=C1)Br)N)Br

Specifications

Melting Point 72°C to 76°C
Quantity 100 g
Beilstein 2085782
Solubility Information Insoluble in water.
Formula Weight 264.96
Percent Purity ≥98%
Chemical Name or Material 2,6-Dibromo-4-methylaniline

RUO – Research Use Only

Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.