missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromo-4-isopropylaniline, 98%

Product Code. 11439190
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439190 5 g 5g
11449190 25 g 25g
11459190 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439190 Supplier Thermo Scientific Alfa Aesar Supplier No. A15932.06
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 10546-65-3 | C9H11Br2N | 293.002 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10546-65-3
Molecular Formula C9H11Br2N
Molecular Weight (g/mol) 293.002
MDL Number MFCD00015475
InChI Key CJEBZUFROMNDEK-UHFFFAOYSA-N
PubChem CID 139184
IUPAC Name 2,6-dibromo-4-propan-2-ylaniline
SMILES CC(C)C1=CC(=C(C(=C1)Br)N)Br

Specifications

Melting Point 45°C to 47°C
Quantity 5 g
Synonym 2,6-dibromo-4-isopropylaniline, 2,6-dibromo-4-methylethyl phenylamine, benzenamine,2,6-dibromo-4-1-methylethyl, benzenamine, 2,6-dibromo-4-1-methylethyl, 2,6-dibromocumidine, 2,6-dibromo-cumidine, acmc-20ao9h, 2,6-dibromo-4-iso-propylaniline, 2,6-dibromo-4-propan-2-yl aniline, 2,6-dibromo-4-1-methylethyl aniline
Formula Weight 293.01
Percent Purity 98%
Chemical Name or Material 2,6-Dibromo-4-isopropylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.