missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,6-Dibromo-4-fluoroaniline, 98%

Product Code. 11480103
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480103 50 g 50g
11470103 10 g 10g
11490103 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480103 Supplier Alfa Aesar™ Supplier No. A12141.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 344-18-3
Molecular Formula C6H4Br2FN
Molecular Weight (g/mol) 268.911
MDL Number MFCD00041445
InChI Key VHPLZFGCNLDYQH-UHFFFAOYSA-N
Synonym 1-amino-2,6-dibromo-4-fluorobenzene, benzenamine, 2,6-dibromo-4-fluoro, pubchem3444, acmc-1cte4, 2,6-dibromo-4-flouroaniline, ksc494q8f, 2,6-dibromo-4-fluoro-aniline, 2,6-dibromo-4-fluorobenzenamine, 2,6-dibromo-4-fluorophenylamine, attercop-chm at130221
PubChem CID 242872
IUPAC Name 2,6-dibromo-4-fluoroaniline
SMILES C1=C(C=C(C(=C1Br)N)Br)F

Specifications

Melting Point 63°C to 66°C
Quantity 50 g
UN Number UN2811
Beilstein 3246923
Formula Weight 268.92
Percent Purity 98%
Chemical Name or Material 2,6-Dibromo-4-fluoroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.