missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromo-4-chlorophenol, TRC

Product Code. 30382294
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382294 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382294 Supplier Toronto Research Chemicals Supplier No. TRCD425818250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5324-13-0
Molecular Formula C6H3Br2ClO
Quantity 250 mg
InChI Formula InChI=1S/C6H3Br2ClO/c7-4-1-3(9)2-5(8)6(4)10/h1-2,10H
SMILES Oc1c(Br)cc(Cl)cc1Br
IUPAC Name 2,6-dibromo-4-chlorophenol
Molecular Weight (g/mol) 286.35
Formula Weight 283.8239
Chemical Name or Material 2,6-Dibromo-4-chlorophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.